Learn More
Hirschmann™ DURAN™ Class AS Micro Burettes Blue Graduations
Product Code. 13158009
Change view
Click to view available options
Capacity (Metric):
2 mL
5 mL
10 mL
Plug Material:
Glass
PTFE
Unit Size:
Pack of 2
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Plug Material unitSize
13158009 10 mL Glass Pack of 2
13138009 2 mL Glass Pack of 2
13168009 2 mL PTFE Pack of 2
13148009 5 mL Glass Pack of 2
13178009 5 mL PTFE Pack of 2
13118009 5 mL Glass Pack of 2
13188009 10 mL PTFE Pack of 2
13128009 10 mL Glass Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 13158009 Supplier Hirschmann™ Supplier No. 3310160
Only null left
Add to basket
Add to basket
Excellent for dispensing measured amounts of chemical solutions in the laboratory. Manufactured from durable DURAN™ glass tubing.
  • Schellbach
  • Bang pattern
  • Main point ring graduation
  • With blue graduation
  • Acc. to DIN EN ISO 385
  • Support base
  • Conformity certified, with dated batch identification
Specifications
Capacity (Metric) 10 mL
Material Borosilicate Glass
Graduations 0.02 mL
Includes Bang, Main Point Ring Graduation, Schellbach Stripes, Socket
Plug Material Glass
Scale Blue
Stopcock Lateral
Stopcock Material Glass
Tolerance ±0.02 mL
Type Microburet
Class Class AS
Certifications/Compliance DIN EN ISO 385