Learn More
Lenz Laborglasinstrumente™ Two-Neck Round Bottom Flasks, Side Neck Angled
Product Code. 10481083
Change view
Click to view available options
Capacity (Metric):
24 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Socket Joint:
14/23 NS Side Neck, 14/23 NS Center Neck
14/23 NS Side Neck, 24/29 NS Center Neck
14/23 NS Side Neck, 29/32 NS Center Neck
19/26 NS Side Neck, 24/29 NS Center Neck
19/26 NS Side Neck, 29/32 NS Center Neck
29/32 NS Side Neck, 29/32 NS Center Neck
NS29/NS14
Unit Size:
Each
Product Code. Capacity (Metric) Socket Joint unitSize
10481083 250 mL 14/23 NS Side Neck, 24/29 NS Center Neck Each
10602662 1000 mL 19/26 NS Side Neck, 24/29 NS Center Neck Each
12633790 100 mL 14/23 NS Side Neck, 14/23 NS Center Neck Each
10332611 100 mL 14/23 NS Side Neck, 29/32 NS Center Neck Each
10070281 100 mL 19/26 NS Side Neck, 29/32 NS Center Neck Each
10372751 250 mL 14/23 NS Side Neck, 29/32 NS Center Neck Each
10204481 250 mL 19/26 NS Side Neck, 29/32 NS Center Neck Each
10284521 250 mL 29/32 NS Side Neck, 29/32 NS Center Neck Each
10382751 500 mL 14/23 NS Side Neck, 29/32 NS Center Neck Each
10388839 50 mL NS29/NS14 Each
10139120 500 mL 29/32 NS Side Neck, 29/32 NS Center Neck Each
10772512 24 mL 14/23 NS Side Neck, 14/23 NS Center Neck Each
10624271 50 mL 14/23 NS Side Neck, 14/23 NS Center Neck Each
10181991 500 mL 19/26 NS Side Neck, 29/32 NS Center Neck Each
14 options
Product Code. 10481083
Brand: Lenz Laborglasinstrumente™ 03213149
Only null left
Add to basket
Add to basket
Especially made for distillation, reactions and to make solutions or hold, contain and collect chemicals. Lenz Laborglasinstrumente™ Two-Neck Round Bottom Flasks, Side Neck Angled are manufactured from durable DURAN™ borosilicate glass.
  • Acc. to DIN 12394
  • With side neck at 20° angle
  • With inscription label
  • Corrosion-free and resistant to thermal shock
  • Ground joint features high taper accuracy and excellent tightness
Specifications
Certifications/Compliance Acc. to DIN 12394 and in addition to DIN standard
Capacity (Metric) 250 mL
Material Borosilicate Glass, Duran Glass
Socket Joint 14/23 NS Side Neck, 24/29 NS Center Neck
Bottom Shape Round