Learn More
Lenz Laborglasinstrumente™ Two-Neck Round Bottom Flasks, Side Neck Angled
Click to view available options
Capacity (Metric):
24 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Socket Joint:
14/23 NS Side Neck, 14/23 NS Center Neck
14/23 NS Side Neck, 24/29 NS Center Neck
14/23 NS Side Neck, 29/32 NS Center Neck
19/26 NS Side Neck, 24/29 NS Center Neck
19/26 NS Side Neck, 29/32 NS Center Neck
29/32 NS Side Neck, 29/32 NS Center Neck
NS29/NS14
Unit Size:
Each
Description
- Acc. to DIN 12394
- With side neck at 20° angle
- With inscription label
- Corrosion-free and resistant to thermal shock
- Ground joint features high taper accuracy and excellent tightness
Specifications
Specifications
| Certifications/Compliance | Acc. to DIN 12394 and in addition to DIN standard |
| Capacity (Metric) | 500 mL |
| Material | Borosilicate Glass, Duran Glass |
| Socket Joint | 19/26 NS Side Neck, 29/32 NS Center Neck |
| Bottom Shape | Round |