Learn More
Lenz Laborglasinstrumente™ Three-Neck Round Bottom Flasks, Side Necks Parallel
Especially made for distillation, reactions and to make solutions or hold, contain and collect chemicals. Lenz Laborglasinstrumente™ Three-Neck Round Bottom Flasks, Side Necks Parallel are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 03305485
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10578081
  • 3853.10 NOK / Each
Estimated Shipment: 02-07-2024
Description
  • Acc. to DIN 12392
  • Two parallel side necks at 90° angle
  • With conical ground joints, featuring high taper accuracy and excellent tightness
  • With inscription label
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
6 L
Borosilicate Glass, Duran Glass
29/32 NS Side Neck, 45/40 NS Center Neck
Round