Learn More
Lenz Laborglasinstrumente™ Three-Neck Round Bottom Flasks, Side Necks Parallel
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
2000 mL
4 L
6 L
10 L
Socket Joint:
14/23 NS Side Neck, 29/32 NS Center Neck
19/26 NS Side Neck, 29/32 NS Center Neck
29/32 NS Side Neck, 29/32 NS Center Neck
29/32 NS Side Neck, 45/40 NS Center Neck
Unit Size:
Each
Description
- Acc. to DIN 12392
- Two parallel side necks at 90° angle
- With conical ground joints, featuring high taper accuracy and excellent tightness
- With inscription label
- Corrosion-free and resistant to thermal shock
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Material | Borosilicate Glass, Duran Glass |
| Socket Joint | 14/23 NS Side Neck, 29/32 NS Center Neck |
| Bottom Shape | Round |