Learn More
Lenz Laborglasinstrumente™ DURAN™ Borosilicate Glass Round Bottom Flasks with Conical Ground Joint
Product Code. 10781352
Change view
Click to view available options
Capacity (Metric):
5 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
3000 mL
5 L
10 L
Socket Joint:
14/23 NS Socket
19/26 NS Socket
24/29 NS Socket
29/32 NS Socket
45/40 NS Socket
Unit Size:
Each
Pack of 10
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Socket Joint unitSize
10781352 5 L 29/32 NS Socket Each
10374971 10 L 29/32 NS Socket Each
10429800 1000 mL 24/29 NS Socket Pack of 10
10171371 1000 mL 45/40 NS Socket Pack of 10
11852552 1000 mL 29/32 NS Socket Pack of 10
15340885 100 mL 14/23 NS Socket Pack of 10
15350895 100 mL 24/29 NS Socket Pack of 10
10627371 100 mL 19/26 NS Socket Pack of 10
10653622 25 mL 19/26 NS Socket Pack of 10
10047830 250 mL 14/23 NS Socket Pack of 10
10772392 250 mL 19/26 NS Socket Pack of 10
10208030 250 mL 24/29 NS Socket Pack of 10
10461083 3000 mL 29/32 NS Socket Each
10697281 50 mL 19/26 NS Socket Pack of 10
10572671 500 mL 24/29 NS Socket Pack of 10
10165350 50 mL 24/29 NS Socket Pack of 10
10529971 5 mL 14/23 NS Socket Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Product Code. 10781352 Supplier Lenz Laborglasinstrumente™ Supplier No. 03002982
  • ONLINE EXCLUSIVE PRICE
    2765.00 NOK
    1868.37 NOK / Pack of 1
    Save 896.63 NOK
    32% Off
Estimated Shipment: 25-08-2026
Specifications
Certifications/Compliance According to DIN EN ISO 4797 and in addition to standard
Capacity (Metric) 5 L
Material Borosilicate Glass, Duran Glass
Socket Joint 29/32 NS Socket
Bottom Shape Round
Diameter (Metric) Outer 223 mm