Learn More
Lenz Laborglasinstrumente™ DURAN™ Borosilicate Glass Round Bottom Flasks with Conical Ground Joint
Click to view available options
Capacity (Metric):
5 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
3000 mL
5 L
10 L
Socket Joint:
14/23 NS Socket
19/26 NS Socket
24/29 NS Socket
29/32 NS Socket
45/40 NS Socket
Unit Size:
Each
Pack of 10
Specifications
Specifications
| Certifications/Compliance | According to DIN EN ISO 4797 and in addition to standard |
| Capacity (Metric) | 25 mL |
| Material | Borosilicate Glass, Duran Glass |
| Socket Joint | 19/26 NS Socket |
| Bottom Shape | Round |
| Diameter (Metric) Outer | 41 mm |