Learn More
- Home
- Products
- Flasks
- Boiling Flasks
- Lenz Laborglasinstrumente™ DURAN™ Borosilicate Glass Round Bottom Flasks with Conical Ground Joint
Lenz Laborglasinstrumente™ DURAN™ Borosilicate Glass Round Bottom Flasks with Conical Ground Joint
Product Code. 10429800
Change view
Click to view available options
Capacity (Metric):
5 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
3000 mL
5 L
10 L
Socket Joint:
14/23 NS Socket
19/26 NS Socket
24/29 NS Socket
29/32 NS Socket
45/40 NS Socket
Unit Size:
Each
Pack of 10
| Product Code. | Capacity (Metric) | Socket Joint | unitSize |
|---|---|---|---|
| 10429800 | 1000 mL | 24/29 NS Socket | Pack of 10 |
| 10374971 | 10 L | 29/32 NS Socket | Each |
| 10171371 | 1000 mL | 45/40 NS Socket | Pack of 10 |
| 11852552 | 1000 mL | 29/32 NS Socket | Pack of 10 |
| 15340885 | 100 mL | 14/23 NS Socket | Pack of 10 |
| 15350895 | 100 mL | 24/29 NS Socket | Pack of 10 |
| 10627371 | 100 mL | 19/26 NS Socket | Pack of 10 |
| 10653622 | 25 mL | 19/26 NS Socket | Pack of 10 |
| 10047830 | 250 mL | 14/23 NS Socket | Pack of 10 |
| 10772392 | 250 mL | 19/26 NS Socket | Pack of 10 |
| 10208030 | 250 mL | 24/29 NS Socket | Pack of 10 |
| 10461083 | 3000 mL | 29/32 NS Socket | Each |
| 10781352 | 5 L | 29/32 NS Socket | Each |
| 10697281 | 50 mL | 19/26 NS Socket | Pack of 10 |
| 10572671 | 500 mL | 24/29 NS Socket | Pack of 10 |
| 10165350 | 50 mL | 24/29 NS Socket | Pack of 10 |
| 10529971 | 5 mL | 14/23 NS Socket | Pack of 10 |
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Specifications
Specifications
| Certifications/Compliance | According to DIN EN ISO 4797 and in addition to standard |
| Capacity (Metric) | 1000 mL |
| Material | Borosilicate Glass, Duran Glass |
| Socket Joint | 24/29 NS Socket |
| Bottom Shape | Round |
| Diameter (Metric) Outer | 131 mm |