Learn More
Lenz Laborglasinstrumente™ DURAN™ Glass Round Bottom Flasks
Can be used to make solutions or hold, contain, collect and measure chemicals. Lenz Laborglasinstrumente™ DURAN™ Glass Round Bottom Flasks are manufactured from durable borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ B0418121
Additional Details : Weight : 0.11100kg
Product Code. 10538460
  • 217.00 NOK / Each
Estimated Shipment: 12-07-2024
Description
  • With inscription label
  • Available in a range of capacities
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
250 mL
Borosilicate Glass, Duran Glass
Head Touches NSH 19/26
Round