Learn More
DWK Life Sciences DURAN™ Blank for separating and dropping funnel, cylindrical with stopper bed for ISO 4800
Product Code. 15231629 Shop All DWK Life Sciences Products
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Pack of 10
Product Code. 15231629
Brand: DWK Life Sciences 210142404
Only null left
Add to basket
Add to basket
Raw body for further processing as dropping funnel.
DURAN™ dropping funnels are used, for example, for the metered supply of liquid to a reaction mixture.
  • Cylindrical with stopper bed for ISO 4800
  • Glass articles for further processing
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric)Top 42 mm
Capacity (Metric) 100 mL
Material Borosilicate Glass (3.3)
Autoclavable Autoclavable
For Use With (Application) Blank for further processing as dropping funnel; Phase separation
Quantity 10/Pk.