Learn More
DWK Life Sciences DURAN™ Blank for reaction vessel
For processing as a reaction vessel.
Brand: DWK Life Sciences 210207602
1444.00 NOK valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 1.72000kg
Description
DURAN™ blanks for reaction vessels are convincing due to their good wall thickness distribution and enable the cost-effective production of reaction vessels with reproducible geometry. For pressure resistance, please refer to our notes in the technical appendix of the laboratory glassware catalogue. For glass articles which deviate from the original form due to further processing, the permissible operating data must be determined in accordance with the technical rules and regulations.
- Component for further processing into reaction vessels
- Due to the geometry and the massive wall thickness, the blanks are suitable for devices with pressure and vacuum applications.
- Glass articles for further processing
Specifications
215 mm | |
Borosilicate Glass (3.3) | |
1/Pk. |
6000 mL | |
For processing as a reaction vessel | |
Yes |