Learn More
DWK Life Sciences DURAN™ Blank for Evaporating Flask, Pear-shape
Product Code. 11649848 Shop All DWK Life Sciences Products
Change view
Click to view available options
Volume (Metric):
100 mL
250 mL
500 mL
1000 mL
2000 mL
3000 mL
Unit Size:
Each
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
11649848 3000 mL Each
15201639 1000 mL Pack of 10
10243401 2000 mL Pack of 10
15281629 250 mL Pack of 10
15291629 500 mL Pack of 10
15271629 100 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11649848 Supplier DWK Life Sciences Supplier No. 210286802
  • ONLINE EXCLUSIVE PRICE
    794.85 NOK
    575.83 NOK / Pack of 1
    Save 219.02 NOK
    27% Off
In Stock
For further processing as ground piston.
DURAN™ evaporating flasks have a uniform wall thickness and good rotational symmetry. For later use in rotary evaporators for the evaporation of solvents, a standard ground joint sleeve is formed or prepared.
  • Pear-shaped
  • Evaporating flasks have a uniform wall thickness and good rotational symmetry
  • Glass articles for further processing
Specifications
Description Autoclavable
Material Borosilicate Glass (3.3)
For Use With (Application) For further processing as ground piston; Rotary evaporator; After further processing it can be used for evaporation of solvents
Quantity 1 Each
Volume (Metric) 3000 mL
Diameter (Metric) 176 mm