Learn More
DWK Life Sciences DURAN™ Blank for Engler Distilling Flask, with beaded rim
For further processing.
Brand: DWK Life Sciences 216512402
Additional Details : Weight : 0.68000kg
Description
Blanks for DURAN™ Engler Distilling Flasks are intended for further processing into DURAN™ Engler flask according to ASTM D86 and DIN EN ISO 3405. Engler flasks are ideal as heating glasses and for distillations due to their uniform wall thickness.
- With beaded rim
- For further processing to DURAN™ Engler flask according to ASTM D86 and DIN EN ISO 3406
- Due to uniform wall thickness ideal as cooking glass and suitable for distillations
- Glass articles for further processing
Specifications
66 mm | |
Borosilicate Glass (3.3) | |
10/Pk. |
100 mL | |
For further processing.; Heating | |
Yes |