Learn More
DWK Life Sciences DURAN™ Bell Jar, with glass knob top
Shop All DWK Life Sciences Products
Click to view available options
Diameter (Metric) Base:
185 mm
260 mm
315 mm
Unit Size:
Each
Description
By default, tests are carried out at normal pressure. The conduct of tests under vacuum (negative pressure) can have significant effects on the reaction sequence. In order to be able to observe these effects in the rough and fine vacuum range, a vacuum experiment plate and a bell jar (also called air pump bell) are required.
- The button makes it easy to raise the bell jar
- Fired-on, highly durable white ceramic
Specifications
Specifications
| Diameter (Metric) Base | 185 mm |
| Material | Borosilicate Glass (3.3) |
| For Use With (Application) | Vacuum applications; Tests in vacuum chambers |
| No. per Pack | 1/Pk. |
| Autoclavable | Autoclavable |