Learn More
DWK Life Sciences DURAN™ Bell Jar, with aperture in neck
Product Code. 10152121 Shop All DWK Life Sciences Products
Change view
Click to view available options
Diameter (Metric) Base:
185 mm
215 mm
315 mm
Unit Size:
Each
Product Code. Diameter (Metric) Base unitSize
10152121 315 mm Each
10727881 185 mm Each
10797481 215 mm Each
3 options
Product Code. 10152121
Brand: DWK Life Sciences 244656903
Only null left
Add to basket
Add to basket
The flat flange bell is used for tests with a vacuum experimental plate (vacuum chamber). The bell has a ground flange and a ground joint for the insertion of measuring equipment.
By default, tests are carried out at normal pressure. The conduct of tests under vacuum (negative pressure) can have significant effects on the reaction sequence. In order to be able to observe these effects in the rough and fine vacuum range, a vacuum experiment plate and a bell jar (also called air pump bell) are required.
  • Tight closure due to standard ground joint
  • Top opening allows connection of additional equipment
  • Fired-on, highly durable white ceramic
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Base 315 mm
Material Borosilicate Glass (3.3)
For Use With (Application) Vacuum applications; Tests in vacuum chambers
No. per Pack 1/Pk.