Learn More
Lenz Laborglasinstrumente™ DURAN™ Beakers
Product Code. p-8005992
Click to view available options
Capacity (Metric):
250 mL
400 mL
1000 mL
2000 mL
Unit Size:
Pack of 10
Product Code. 11923621
Brand: Lenz Laborglasinstrumente™ 07004049
Only null left
Add to basket
Add to basket
Ideal for heating applications in the laboratory. Lenz Laborglasinstrumente™ DURAN™ Beakers feature an easy-to-read scale and a large labeling area in fired-on, durable white ceramic.
  • Spout for clean pouring
  • Uniform wall thickness distribution
  • Meets DIN 12331/ISO 3819 standards
  • Constructed from DURAN™ borosilicate glass
Specifications
Diameter (Metric) Outer 60mm
Capacity (Metric) 250 mL
Material Borosilicate Glass
Height (Metric) 120mm
Quantity 10Pack
Type Beaker
Graduated Yes
Certifications/Compliance DIN 12331/ISO 3819