Learn More
DWK Life Sciences DURAN™ Aspirator Laboratory Bottle, Unground Neck for Rubber Stopper, Side Arm for attaching Tubing
Product Code. 15595016 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
Unit Size:
Each
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15595016 1000 mL Each
15281999 250 mL Pack of 10
15291999 500 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15595016 Supplier DWK Life Sciences Supplier No. 247085407
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Aspirator bottle with side-arm
Simple side-arm aspirator bottles. Fitted with an integral side-arm for the attachment of 9 mm bore flexible tubing, these simple aspirator bottles are available in capacities up to 1 liter. Unground neck to accept rubber stoppers.
  • Outlet facilitates attachment of flexible tubing
  • With plain neck and bottom side arm
  • Made of virtually inert, chemically resistant DURAN™ glass
Specifications
Capacity (Metric) 1000 mL
For Use With (Application) Storage and dispensing of liquids and reagents
Material Borosilicate Glass (3.3)
No. per Pack 1/Pk.
Diameter (Metric) 111 mm