Learn More
DWK Life Sciences DURAN™ Aspirator Laboratory Bottle, Unground Neck for Rubber Stopper, Side Arm for attaching Tubing
Shop All DWK Life Sciences ProductsDescription
Simple side-arm aspirator bottles. Fitted with an integral side-arm for the attachment of 9 mm bore flexible tubing, these simple aspirator bottles are available in capacities up to 1 liter. Unground neck to accept rubber stoppers.
- Outlet facilitates attachment of flexible tubing
- With plain neck and bottom side arm
- Made of virtually inert, chemically resistant DURAN™ glass
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| For Use With (Application) | Storage and dispensing of liquids and reagents |
| Material | Borosilicate Glass (3.3) |
| No. per Pack | 10/Pk. |
| Diameter (Metric) | 89 mm |
| Autoclavable | Autoclavable |