Learn More
Lenz Laborglasinstrumente™ Duran™ Allihn Condensers with Cone
Specifications
Specifications
| For Use With (Application) | For Condensing Vapors |
| Includes | DURAN™ Olive Tubing |
| Quantity | 1 Pack |
| Standard Taper Joint No | NS 45/40 |
| Material | Duran Glass |
| Type | Allihn Condenser |