Learn More
DWK Life Sciences DURAN™ Adapter, standard ground joint
Product Code. 10757101 Shop All DWK Life Sciences Products
Change view
Click to view available options
Type:
Expansion Joint
Expansion Joint, Non-DIN Size
Reduction Joint
Reduction Joint, Non-DIN Size
Joint Size:
Socket NS 14/23, Cone NS 19/26
Socket NS 14/23, Cone NS 24/29
Socket NS 14/23, Cone NS 29/32
Socket NS 19/26, Cone NS 14/23
Socket NS 19/26, Cone NS 24/29
Socket NS 19/26, Cone NS 29/32
Socket NS 24/29, Cone NS 14/23
Socket NS 24/29, Cone NS 19/26
Socket NS 24/29, Cone NS 29/32
Socket NS 29/32, Cone NS 14/23
Socket NS 29/32, Cone NS 19/26
Socket NS 29/32, Cone NS 24/29
Unit Size:
Each
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Joint Size unitSize
10757101 Reduction Joint Socket NS 19/26, Cone NS 14/23 Each
10788251 Expansion Joint Socket NS 14/23, Cone NS 29/32 Each
10499722 Expansion Joint, Non-DIN Size Socket NS 14/23, Cone NS 24/29 Each
10171401 Expansion Joint Socket NS 19/26, Cone NS 29/32 Each
10029840 Reduction Joint, Non-DIN Size Socket NS 24/29, Cone NS 14/23 Each
10622452 Expansion Joint, Non-DIN Size Socket NS 24/29, Cone NS 29/32 Each
10314961 Reduction Joint Socket NS 29/32, Cone NS 14/23 Each
10182021 Expansion Joint Socket NS 14/23, Cone NS 19/26 Each
10622642 Reduction Joint Socket NS 29/32, Cone NS 19/26 Each
10203601 Expansion Joint, Non-DIN Size Socket NS 19/26, Cone NS 24/29 Each
10039840 Reduction Joint, Non-DIN Size Socket NS 24/29, Cone NS 19/26 Each
10151451 Reduction Joint, Non-DIN Size Socket NS 29/32, Cone NS 24/29 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Product Code. 10757101 Supplier DWK Life Sciences Supplier No. 241142601
Only null left
Add to basket
Add to basket
The transition adapter connects two glass components with different standard ground joints.
Transition adapters or reducers are glass components with two standard ground joints of different sizes. With the help of these adapters, glass components with different standard ground joints can be connected to each other.
  • Tight closure due to standard ground joint
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Ea.
Item Description Connection of two glass components
Type Reduction Joint
Joint Size Socket NS 19/26, Cone NS 14/23