Learn More
DWK Life Sciences DURAN™ Adapter, standard ground joint
Shop All DWK Life Sciences Products
Click to view available options
Type:
Expansion Joint
Expansion Joint, Non-DIN Size
Reduction Joint
Reduction Joint, Non-DIN Size
Joint Size:
Socket NS 14/23, Cone NS 19/26
Socket NS 14/23, Cone NS 24/29
Socket NS 14/23, Cone NS 29/32
Socket NS 19/26, Cone NS 14/23
Socket NS 19/26, Cone NS 24/29
Socket NS 19/26, Cone NS 29/32
Socket NS 24/29, Cone NS 14/23
Socket NS 24/29, Cone NS 19/26
Socket NS 24/29, Cone NS 29/32
Socket NS 29/32, Cone NS 14/23
Socket NS 29/32, Cone NS 19/26
Socket NS 29/32, Cone NS 24/29
Unit Size:
Each
Description
Transition adapters or reducers are glass components with two standard ground joints of different sizes. With the help of these adapters, glass components with different standard ground joints can be connected to each other.
- Tight closure due to standard ground joint
Specifications
Specifications
| Quantity | 1 Ea. |
| Item Description | Connection of two glass components |
| Type | Reduction Joint, Non-DIN Size |
| Joint Size | Socket NS 29/32, Cone NS 24/29 |