Learn More
DuraCal™ 4.01 Value pH Buffer, Hamilton™
Product Code. 10214122
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10214122 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10214122 Supplier Hamilton Supplier No. 238217
  • ONLINE EXCLUSIVE PRICE
    603.76 NOK
    375.33 NOK / 500mL
    Save 228.43 NOK
    37% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
Economical choice since only about 20mL of buffer is used per calibration. Hamilton™ DuraCal™ 4.01 Value pH Buffers have actual values determined by a DKD laboratory, accredited for pH measurement.
  • 4.01 pH value
  • Immune to micro-organisms
  • Expiration date on the bottle
  • First class certificate with traceability to international standards
Specifications
pH 4.01
Quantity 500 mL