Learn More
H-B Instrument™ Durac™ Timers
Product Code. p-4604597 Shop All SP Scienceware Products
Click to view available options
Time Range:
23 hr. 59 min. 59 sec.
60 min.
99 hr. 59 min. 59 sec.
99 min. 59 sec.
No. of Channels:
1
4
Height (Metric):
3.14 cm
5.1 cm
5.7 cm
7 cm
Width (Metric):
5.7 cm
6.7 cm
7 cm
8 cm
Unit Size:
Each
Product Code. 17673125
Brand: H-B Instrument™ B617003000
Only null left
Add to basket
Add to basket
Includes
Multi-language instructions (English, French, German, Italian, Portuguese and Spanish)
Available in variety of styles
  • Individually tested
  • Model 511 features switchable timings in hour:min. or min.:sec.
Certifications
CE marked, NIST traceable, DAkkS, Meets RoHS specifications.
Specifications
Product Type Timer
Time Range 23 hr. 59 min. 59 sec.
Accuracy 3 sec.
Display Digital
No. of Channels 1
Battery Type LR44
Width (English) 2.8 in.
Width (Metric) 7 cm
Height (English) 2 in.
Height (Metric) 5.1 cm
Includes Memory, clock, multi language instructions (English, French, German, Italian, Portuguese and Spanish) Statement of Traceability indicating accuracy traceable to NIST and DAkkS
Certifications/Compliance CE, ROHS, NIST, DAkkS