Learn More
H-B Instrument™ Durac™ Barometer
Product Code. p-4604540 Shop All SP Scienceware Products
Change view
Click to view available options
Diameter (Metric) Face:
64 mm
Material:
Plastic
Model:
3938
Unit Size:
Each
Product Code. Diameter (Metric) Face Material Model unitSize
17580685 64 mm Plastic 3938 Each
1 options
Product Code. 17580685
Brand: H-B Instrument™ B615020300
Only null left
Add to basket
Add to basket
Recalibrateable unit
  • Features black lines and numbers
  • Hole present for wall mounting
  • Adjust automatically once set
  • Designed to work in altitudes below 6562 ft. (2000m)
Specifications
Model 3938
Type Barometer
Length (English) 2.5 in.
Length (Metric) 64 mm
Material Plastic
Diameter (Metric) Face 64 mm