Learn More
Keystone Cleanroom Products DuPont™ Tyvek™ Autoclave Pouch with Steam Indicator Dot
GREENER_CHOICE
Product Code. 17748247 Shop All Keystone Cleanroom Products Products
Change view
Click to view available options
Dimensions (L x W):
12 in. x 10 in.
12 x 8 in.
14 in. x 8 in.
16 in. x 6 in.
18 in. x 12 in.
20 in. x 10 in.
20 in. x 20 in.
22 in. x 14 in.
22 in. x 30 in.
22 in. x 6 in.
24 in. x 18 in.
25 in. x 10 in.
25 in. x 5 in.
28 in. x 14 in.
28 in. x 26 in.
28 in. x 48 in.
30 in. x 18 in.
30 in. x 8 in.
32 in. x 12 in.
32 in. x 20 in.
34 in. x 34 in.
34 in. x 6 in.
36 in. x 16 in.
36 in. x 3 in.
37 in. x 25 in.
40 in. x 18 in.
40 in. x 34 in.
44 in. x 16 in.
44 in. x 24 in.
68 in. x 18 in.
8 in. x 6 in.
9 in. x 8 in.
Unit Size:
Pack of 100
Pack of 50
Product Code. Dimensions (L x W) unitSize
17748247 40 in. x 18 in. Pack of 50
17708267 8 in. x 6 in. Pack of 100
17748267 9 in. x 8 in. Pack of 100
17718267 12 x 8 in. Pack of 100
17738237 12 in. x 10 in. Pack of 100
17728267 14 in. x 8 in. Pack of 100
17778257 16 in. x 6 in. Pack of 100
17768237 18 in. x 12 in. Pack of 100
17748237 20 in. x 10 in. Pack of 100
17768247 20 in. x 20 in. Pack of 50
17788257 22 in. x 6 in. Pack of 50
17788237 22 in. x 14 in. Pack of 100
17718257 22 in. x 30 in. Pack of 50
17728247 24 in. x 18 in. Pack of 50
17768257 25 in. x 5 in. Pack of 100
17758237 25 in. x 10 in. Pack of 50
17798237 28 in. x 14 in. Pack of 100
17708257 28 in. x 26 in. Pack of 50
17758257 28 in. x 48 in. Pack of 50
17738267 30 in. x 8 in. Pack of 50
17738247 30 in. x 18 in. Pack of 50
17778237 32 in. x 12 in. Pack of 100
17778247 32 in. x 20 in. Pack of 50
17798257 34 in. x 6 in. Pack of 100
17728257 34 in. x 34 in. Pack of 50
17748257 36 in. x 3 in. Pack of 50
17708247 36 in. x 16 in. Pack of 50
17798247 37 in. x 25 in. Pack of 50
17738257 40 in. x 34 in. Pack of 50
17718247 44 in. x 16 in. Pack of 50
17788247 44 in. x 24 in. Pack of 50
17758247 68 in. x 18 in. Pack of 50
32 options
Product Code. 17748247
Brand: Keystone Cleanroom Products COVMGTVKUB18X40
Only null left
Add to basket
Add to basket
Manufactured in a ISO6 cleanroom to ensure cleanliness.
  • Designed for steam sterilization of pharmaceutical production equipment and components
  • 1073B Tyvek™ construction allows for optimal steam penetration during sterilization
  • Excellent impact resistance
  • Consistently strong factory seals
  • Exceptional microbial barrier
  • Resists tearing and punctures from sharp corners and heavy machine parts
  • Processed and packaged in a controlled cleanroom environment
  • Lot traceable
Specifications
Autoclavable Autoclavable
Capacity (English) Application Dependent
Certifications/Compliance Certificate of Conformance (COC)
Closure Type Heat Seal
Color White
Disposable Single-use
For Use With (Application) Steam Sterilization
For Use With (Equipment) Pharmaceutical Equipment Change Parts/Components (Stoppers, Seals, etc.)
Material Tyvek
Sterility Non-sterile
Latex Free Yes
Moisture Vapor Transmission Rate 1615 g/m2/24 hr
No. per Case 50/Cs.
Sterilization Method Steam
Dimensions (L x W) 40 in. x 18 in.
Length (English) 40 in.
Length (Metric) 101.6 cm
Width (English) 18 in.
Width (Metric) 45.7 cm
Height (English) 40 in.
Product Type Autoclave Bag