missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUOX2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
2690.00 NOK - 5155.00 NOK
Specifications
| Antigen | DUOX2 |
|---|---|
| Concentration | 1.2 mg/mL |
| Dilution | Western Blot 2 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin reported in scientific literature (PMID 24492313), KnockDown Validated |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18760934
|
Novus Biologicals
NB110-61576SS |
0.025 mL |
2690.00 NOK
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18798573
|
Novus Biologicals
NB110-61576 |
0.1 mL |
5155.00 NOK
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DUOX2 Polyclonal specifically detects DUOX2 in Human, Mouse, Rat, Porcine, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| DUOX2 | |
| Western Blot 2 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Paraffin reported in scientific literature (PMID 24492313), KnockDown Validated | |
| Polyclonal | |
| Rabbit | |
| Cell Biology, Lipid and Metabolism, Neuroscience | |
| PBS and 30% Glycerol with 0.1% Sodium Azide | |
| dual oxidase 2, dual oxidase-like domains 2, EC 1.11.1.-, EC 1.6.3.1, flavoprotein NADPH oxidase, Large NOX 2, LNOX2P138(TOX), Long NOX 2, NADH/NADPH thyroid oxidase p138-tox, NADPH oxidase/peroxidase DUOX2, NADPH thyroid oxidase 2, nicotinamide adenine dinucleotide phosphate oxidase, NOXEF2, p138 thyroid oxidase, P138-TOX, THOX2TDH6, Thyroid oxidase 2 | |
| DUOX2 | |
| IgG | |
| Affinity Purified |
| 1.2 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Pig, Canine | |
| Q9NRD8 | |
| 50506 | |
| Synthetic peptide made to an internal portion of human DUOX2 (within residues 400-500). [Swiss-Prot# Q9NRD8] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title