Description
Featuring two indicator zones for more accurate readings. Macherey Nagel™ Duotest™ pH Paper Rolls give the user the best possible accuracy in pH measurement.
- Provides rapid and reliable results
- Available for a range of pH gradations
- Shows two different colours for each pH-value at intervals of 0.3 to 1 pH unit
- Allows more accurate reading and good estimation of intermediate values
Specifications
Specifications
| pH Range | 9.5 to 14 |
| Format | Roll |
| Graduations | 9.5, 10.0, 10.5, 11.0, 11.5, 12.0, 12.5, 13.0, 13.5, 14.0 |
| Quantity | 1 Each |
| Type | pH Paper |
| Length (Metric) | 5 m |
| Width (Metric) | 10 mm |