Learn More
Macherey-Nagel™ Duotest™ pH Paper Refill
Shop All Macherey-Nagel Products
Click to view available options
pH Range:
1 to 12
1 to 4.3
3.5 to 6.8
5 to 8
7 to 10
Unit Size:
Pack of 2
Pack of 3
Description
- Provides rapid and reliable results
- Available for a range of pH gradations
- Shows two different colors for each pH-value at intervals of 0.3 to 1 pH unit
- Allows more accurate reading and good estimation of intermediate values
Specifications
Specifications
| pH Range | 5 to 8 |
| Format | Refill |
| Quantity | 3 Pack |
| Type | pH Paper |