Learn More
Euro Protection™ Dunlop™ Hygrade Boots
Product Code. 12004475
Change view
Click to view available options
Size:
38
39
40
41
42
43
44
45
46
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12004475 41 Each
12084465 39 Each
12074465 38 Each
12044475 45 Each
15743109 43 Each
12034475 44 Each
12094465 40 Each
12014475 42 Each
12054475 46 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 12004475 Supplier Euro Protection™ Supplier No. 9HYGR41
  • ONLINE EXCLUSIVE PRICE
    643.65 NOK
    281.20 NOK / Pack of 1
    Save 362.45 NOK
    56% Off
Estimated Shipment: 04-09-2026
Specifications
Certifications/Compliance CE Certified, EN ISO 20347
Color White, Green
Footbed Material PVC
For Use With (Application) For industrial use, food companies and agricultural work
Liner Material Nylon
Material PVC
Size 41
Sole Features Anti-slip
Sole Material PVC
Style Non-safety
Resistant To Water
Waterproof Yes
Product Type Boots