Learn More
Thermo Scientific™ DualSpin Rotor Body
Product Code. 15580131 Shop All Thermo Scientific Products
Change view
Click to view available options
:
DualSpin Rotor Body
Product Code.
15580131 DualSpin Rotor Body
1 options
Product Code. 15580131
Brand: Thermo Scientific™ 75008813
Run common clinical IVD blood tubes, gel tubes, 10 mL standard syringes and 15 mL conical tubes with the Thermo Scientific™ DualSpin™ rotor.
TRUSTED_SUSTAINABILITY
Specifications
Material Lightweight Composite
Type Rotor Body
For Use With (Equipment) accuSpin™ 8C Benchtop Centrifuge