Learn More
VEE GEE Dual Scale Hydrometer
Click to view available options
Baume Range:
0° to 26° (Heavy)
0° to 41° (Heavy)
0° to 45° (Heavy)
0° to 72° (Heavy)
10° to 100° (Light)
24° to 43 (Heavy)
25° to 42° (Heavy)
42° to 55° (Heavy)
55° to 65° (Heavy)
64° to 72° (Heavy)
70° to 10° (Light)
Specific Gravity Range:
0.600 to 1.000
0.70 to 1.00
1.000 to 1.220
1.000 to 1.225
1.000 to 1.400
1.000 to 1.450
1.000 to 2.000
1.200 to 1.420
1.200 to 1.425
1.400 to 1.620
1.600 to 1.820
1.800 to 2.020
Conditionnement:
Each
Description
- The Dual Scale Specific Gravity/Baume Hydrometer includes two scales.
- One scale is designed for liquids heavier than water.
- The other scale is specifically for liquids lighter than water.
Spécification
Spécification
| Type | Specific Gravity and Baume Hydrometer |
| Increments | 0.005/1° |
| Length (Metric) | 16.5 cm |
| Baume Range | 24° to 43 (Heavy) |
| Specific Gravity Range | 1.200 to 1.425 |
| Temperature of Standardization | 15.6°C (60°F) |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu