Learn More
Jenway™ Dual Cell Holder
Description
- For use exclusively with Model 6320D Visible Spectrophotometer
- Holds 10mm square cuvettes and 12.7mm diameter tubes
Specifications
Specifications
| Type | Dual Cell Holder |
| For Use With (Equipment) | Jenway 6320D Visible Spectrophotometer |
| Holds | 2 x 10 mm cuvettes and 12.7 mm dia tubes |