Learn More
Fisherbrand™ Anodized Dual Aluminum Blocks
Product Code. 11401997
Click to view available options
Product Type:
D1196, 96-Well
D12384, 384 WELL PCR PLATE
Unit Size:
Each
Product Code. 11401997
Brand: Fisherbrand™ D1296
Useful for a variety of applications in molecular biology, histology, clinical, environmental and industrial. Fisherbrand™ Anodized Dual Aluminum Blocks are for use with Fisherbrand Digital Block Heaters.
  • Excellent heat transfer
  • Anodized to resist thermal shock
  • For holding test tubes for digital heaters
  • Available to fit a range of tube counts and sizes
Specifications
For Use With (Equipment) Micro plate or 4 Slides.
Product Type D1196, 96-Well
Quantity 1 CT