missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DTX4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25222-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
DTX4 is a regulator of Notch signaling, a signaling pathway involved in cell-cell communications that regulates a broad spectrum of cell-fate determinations. It functions as an ubiquitin ligase protein in vivo, mediating ′Lys48′-linked polyubiquitination and promoting degradation of TBK1, targeting to TBK1 requires interaction with NLRP4.Specifications
DTX4 | |
Polyclonal | |
Unconjugated | |
DTX4 | |
deltex homolog 4 (Drosophila), Deltex4, DTX4, KIAA0937, Protein deltex 4, RING finger protein 155, RNF155 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
207521, 23220 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunocytochemistry | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6PDK8, Q9Y2E6 | |
Dtx4 | |
DTX4 Fusion Protein Ag18415 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |