Learn More
IKA™ DT-50-M-Gamma Tubes with Rotor Stator Element
Shop All IKA Products
Click to view available options
For Use With (Equipment):
DT-50-M
Unit Size:
Pack of 10
Specifications
Specifications
| Product Type | DT-50-M Tube |
| Volume Range (Metric) | 15 to 50 mL |
| For Use With (Application) | Dispersion, homogenization, suspensions |
| For Use With (Equipment) | DT-50-M |
| Packaging Quantity | 10/Pk. |