Learn More
Applied Biosystems™ DS-33 Matrix Standard Kit Dx (Dye Set G5)
Product Code. 15998826 Shop All Applied Biosystems Products
Change view
Click to view available options
Description:
Dx DS-33 Matrix Standard Kit, 8 runs, LIZ™ Dye, Wet Ice, For 3500 Dx Genetic Analyzer, 2 to 8°C Storage Temperature
Unit Size:
8 tests
Product Code. Description unitSize
15998826 Dx DS-33 Matrix Standard Kit, 8 runs, LIZ™ Dye, Wet Ice, For 3500 Dx Genetic Analyzer, 2 to 8°C Storage Temperature 8 tests
1 options
Product Code. 15998826
Brand: Applied Biosystems™ a25775
Includes
Contains 5-dye labeled oligonucleotides used for generating the multicomponent matrix in a 1X TE solution
Software for these instruments uses the multicomponent matrix to automatically analyze the five different colored fluorescent dye-labeled samples.
The DS-33 Matrix Standard Kit Dx (Dye Set G5) is used to generate the 'multicomponent matrix' required when analyzing 6-FAM™, VIC™, NED™, PET™, and LIZ™ dye-labeled DNA fragments on the Applied Biosystems™ 3500 Dx Series Genetic Analyzers.
Order Info
Each
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Contains 5-dye labeled oligonucleotides used for generating the multicomponent matrix in a 1X TE solution. Store at 2 to 8°C
Description Dx DS-33 Matrix Standard Kit, 8 runs, LIZ™ Dye, Wet Ice, For 3500 Dx Genetic Analyzer, 2 to 8°C Storage Temperature
For Use With (Equipment) 3500 Dx Genetic Analyzer
Label or Dye LIZ
Product Type DS-33 Matrix Standard Kit
Quantity 8 runs
For research use only