Learn More
Thermo Scientific™ Dry-Bags™ Reusable Filter
Filter water coming into Thermo Scientific™ Dry-Bags™ with a reusable filter that can be used up to 10 times if autoclaved between each use.
Brand: Thermo Scientific™ DB0200A
Additional Details : Weight : 0.01000kg
Description
Remove the need to pre-sterilize water prior to filling Thermo Scientific™ Dry-Bags™ using a sterile, reusable, 0.2 micron Thermo Scientific Dry-Bags Reusable Filter.
Obtain up to 10 uses from the Dry-Bags Reusable Filter, if autoclaved at 121ºC for 15 minutes between each use.Specifications
Dry-Bags Reusable Filter | |
0.2 μm | |
Includes reusable filter only | |
0.050 kg |
2.5 L/min. | |
Dry-Bags | |
Dry-Bags Reusable Filter |