Learn More
Thermo Scientific™ Dry-Bags™ Buffered Peptone Water (ISO)
Product Code. 11419858
Change view
Click to view available options
Format:
Dry-Bags, without filter
Yield:
20L
Packungsgröße:
Pack of 10
Artikelnummer. Format Yield unitSize
11419858 Dry-Bags, without filter 20L Pack of 10
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11419858
Marke: Thermo Scientific™ DB1049W
Edge
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Simplify media preparation in food laboratories with this high quality dehydrated medium in a convienient bag format; just open-fill-dispense, saving precious time in the lab.
Save time in preparing bulk media and diluents for food microbiology testing using Thermo Scientific™ Dry-Bags™ Buffered Peptone Water (ISO). The irradiated media is supplied in lightweight, transparent plastic bags — just add water and the medium is ready for use. Each bag makes 20L of medium, which can be dispensed through the outlet tube.
Use Buffered Peptone Water (ISO) as a non-selective pre-enrichment medium for the isolation of Salmonella species from food and associated samples. As Salmonella may be present in low numbers and/or sub-lethally injured, pre-enrichment allows the cells time to repair and multiply before being introduced to selective culture, thereby improving the chances of recovery from the sample.
Convenient
  • Quick and easy to prepare – just add deionized water through filter sterilization to each Dry-Bag, and they are ready to use in minutes. Pre-sterilized water can also be added under aseptic conditions.
Cost-Effective
  • Light to transport – there is no weighing of media or sterilization required, reducing the burden on media preparation departments and equipment.
Safe
  • No handling of glassware or weighing of culture media required, no need to add supplementary reagents — empty bags can be disposed of with normal laboratory waste.
Quality Guaranteed
  • Quality certificates are available for every batch to help reduce the burden of QC testing.
Workflow
  • Dry-Bags are easily adaptable to laboratory workflows and equipment such as automated diluters.
Easy-To-Use
  • Dry-Bags are easy to fill and dispense with minimum training.
Space-Saving
  • Conveniently packed in flat boxes of five or ten, Dry-Bags take up relatively little space compared to prepared media and can be made up as required.
Re-hydrated Dry-Bags medium can be stored for 3 days at room temp (away from strong light sources) and are available with filters attached for convenience or without filters as a lower-cost option. For those Dry-Bags supplied without filters, autoclavable, reusable filters are also available (DB0200A). For added convenience, it is recommended that the Dry-Bags be filled using the Thermo Scientific Peristaltic Pump (DB2000A or DB3000A) and dispensed using the Thermo Scientific Automated Diluter (DB4000A).
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Spezifikation
Color Clear, colorless - straw
Content And Storage 10°C to 30°C
Description Dry-Bags Buffered Peptone Water (ISO)
Format Dry-Bags, without filter
Product Type Pre-enrichment Medium
Quantity 10/Pk.
Yield 20L