Learn More
Lenz Laborglasinstrumente™ Dropping funnel, conical-shaped with glass plug
Product Code. 11886144
Change view
Click to view available options
Capacity (Metric):
50 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
11886144 2000 mL Each
11876074 50 mL Each
11866124 500 mL Each
11876114 1000 mL Each
4 options
Product Code. 11886144
Brand: Lenz Laborglasinstrumente™ 04457304
Only null left
Add to basket
Add to basket
Made from borosilicate glass. Lenz Laborglasinstrumente™ Dropping funnel, conical-shaped with glass plug is used in the laboratory to transfer fluids. The funnel is fitted with a stopcock which allows the flow to be controlled.
  • Conical shape with NS socket and NS cone
  • With solid glass plug, with screwthread retaining device
  • With inscription label, without scale
  • Borosilicate glass
Specifications
Capacity (Metric) 2000 mL
Material Borosilicate Glass
Pore Size 6.0mm
Quantity 1
Length (Metric) Overall 100 mm