Learn More
Thermo Scientific™ μDrop™ and μDrop Duo Plates
Product Code. p-9177001 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
μDrop Duo Plate
μDrop Plate
Unit Size:
Each
Product Code. Product Type unitSize
16638987 μDrop Duo Plate Each
11899143 μDrop Plate Each
2 options
Product Code. 16638987
Brand: Thermo Scientific™ N12391M2
Thermo Scientific™ μDrop and μDrop Duo plates are used for low volume (2-10 μl) photometric DNA, RNA and protein quantitation and purity analysis. The μDrop plates are compatible with Thermo Scientific Varioskan LUX and Multiskan SkyHigh microplate readers. The ready-made protocols in SkanIt software or Multiskan SkyHigh user interface make quantification fast and easy.
Thermo Scientific™ μDrop™ and μDrop™ Duo Plates are ideal tools for quick and easy photometric low-volume (2–10 μl) concentration measurements of DNA, RNA, protein samples, or spectral scanning. The low volume area is designed to be used with 8-channel pipette for fast and accurate multiple sample pipetting. The plates also contain a separate area for cuvette measurement. μDrop Plates are compatible with Thermo Scientific Varioskan™ LUX, Multiskan™ Sky and Multiskan SkyHigh microplate readers. The intuitive Thermo Scientific SkanIt™ software and Multiskan SkyHigh user interface offer ready-made measurement protocols for quick sample measurement and analysis.
  • dsDNA detection range from a few micrograms to a few milligrams per milliliter
  • Up to 16 (μDrop Plate) or 32 (μDrop Duo Plate) sample positions for quick and easy measurement of sample volumes down to 2 μl
  • Compatible with an eight-channel pipette for sample addition
  • Easy to clean after measurement
  • Measurement slot for a rectangular cuvette
  • Ready-made Multiskan SkyHigh user interface and SkanIt Software protocols for nucleic acid and protein analysis
  • Compatible with Multiskan Sky, Multiskan SkyHigh and Varioskan LUX microplate readers
TRUSTED_SUSTAINABILITY
Specifications
Product Type μDrop Duo Plate
For Use With (Equipment) Varioskan LUX, Multiskan Sky, Multiskan SkyHigh
Plate Type μDrop Duo Plate
Capacity (Metric) 2 to 10μL (low-volume function)
For Use With (Application) Nucleic acid and protein concentration measurement
No. of Positions 32