Learn More
Buerkle™ Drop-boy Dropping Bottles
Product Code. 10056901 Shop All Bürkle Products
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10056901 50 mL Each
10438831 10 mL Each
10445951 25 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10056901 Supplier Buerkle™ Supplier No. 03070050
  • ONLINE EXCLUSIVE PRICE
    19.00 NOK
    17.34 NOK / Pack of 1
    Save 1.66 NOK
    8% Off
View all promotions
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
Use Buerkle™ Drop-boy Dropping Bottles for accurate, reliable and repeatable dispensing of reagents.
  • HDPE
  • Drop dispenser and separate screw cap
  • Closure included
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 50 mL
Color White
Diameter (English) 1.57 in.
Diameter (Metric) Inner Neck 13 mm
Includes Drop dispenser and separate screw cap
Height (Metric) With Closure 113 mm
Material HDPE
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 77 mm
Height (English) 3.03 in.
Diameter (Metric) 40 mm
Closure Type Drip Closure
Autoclavable Not Autoclavable
Capacity (English) 1.69 oz.
Shape Round
Disposable Reusable