Learn More
Buerkle™ Drill Bit for Buerkle™ Mole
Shop All Bürkle Products
Click to view available options
For Use With (Application):
For Clay and Wet Soils
For Coarse Sand
For Collecting and Removing Stones
For Combination Soils
For Gravel
For Riverbank Soil Horizons
For Sand
Unit Size:
Each
Description
- Clay and damp soil
- With narrow side panels for minimum soil friction/resistance. As wet soils are usually very cohesive, the soil still adheres to the bit very well.
- Sand
- With broad side panels and therefore a large retention capacity for the sand.
- Coarse sand
- With extra broad side panels enclosing even more of the bit so that even coarse sandy soils and very dry soils are retained.
- Combination
- Retains both strongly cohesive and less cohesive soils.
- Riverside
- Very suitable for drilling in hard, encrusted soils and soils mixed with fine gravel. The drill cutters project from the drill itself, so that friction is greatly reduced.
- Gravel drill
- With slightly rounded, oblique scoops which pull themselves downward in the gravel. Any stones present are clamped between the drill sides. Finer soil material (coarse sand with fine gravel) is retained in the drill bit.
- Stone catcher
- The stone catcher is used to collect and remove stones that prevent proper sampling with one of the other drill bits.
Specifications
Specifications
| Product Type | Drill Bit |
| Description | Equipment for mole drill sampler |
| For Use With (Application) | For Collecting and Removing Stones |
| For Use With (Equipment) | Mole Drill Sampler |