Learn More
Lenz Laborglasintrumente™ Drechsel Gas Washing Bottle without Filter Disc
Product Code. 10391760
Change view
Click to view available options
Capacity (Metric):
250 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10391760 250 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10391760 Supplier Lenz Laborglasintrumente™ Supplier No. 05500512
Only null left
Add to basket
Add to basket
Gas washing bottles acc. to Drechsel, without filter disc, complete with head
  • Acc. to DIN 12596
  • Complete with head
  • Ground joint connection: NS 29/32
  • Material: DURAN™ tubing (head), borosilicate glass 3.3 (bottle)
Specifications
Capacity (Metric) 250 mL
Type Thread GL 14
Quantity 1