Learn More
Thermo Scientific™ DreamTaq Green PCR Master Mix (2X) Promotion
Product Code. 15717526 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
4 x 1.25 mL
20 x 1.25mL
No. of Reactions:
200
1000
Unit Size:
1000 reactions
200 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity No. of Reactions unitSize
15717526 20 x 1.25mL 1000 1000 reactions
15707526 4 x 1.25 mL 200 200 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15717526 Supplier Thermo Scientific™ Supplier No. K1082
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Contains green buffer that includes density reagent and two tracking dyes for direct loading of PCR products on gels
  • DreamTaq DNA Pol,
  • dNTPs,
  • Green buffer, density reagent and dyes for direct loading,
  • supplied with Water,
  • Nuclease free
TRUSTED_SUSTAINABILITY
Specifications
Concentration 2X
Quantity 20 x 1.25mL
For Use With (Application) Standard PCR, RT-PCR, Genotyping, Cloning
No. of Reactions 1000