Learn More
Labconco™ Drain Hose Plug
Product Code. 17285494
Change view
Click to view available options
Capacity (Metric):
Up to 18 L
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
17285494 Up to 18 L Each
1 options
Product Code. 17285494
Brand: Labconco™ 7728000
Only null left
Add to basket
Add to basket
Drain plug fits FreeZone Freeze dryers with retractable drain hose.
  • Drain plug fits FreeZone Freeze dryers with retractable drain hose.
  • Not compatible with models created after 2005.
Specifications
Type Stopper
Capacity (Metric) Up to 18 L
Temperature Range (Metric) -15°C to +30°C
Material Polypropylene
Height (English) 1.37 in.
Height (Metric) 34.8 mm
Length (English) 1.38 in.
Length (Metric) 35 mm