Learn More
Cytiva Dracon sponge, thickness = 6 mm (0.25 inch)
Product Code. 11500914 Shop All Cytiva Products
Change view
Click to view available options
Thickness (Metric):
0.6 cm
Unit Size:
Pack of 3
Product Code. Thickness (Metric) unitSize
11500914 0.6 cm Pack of 3
1 options
Product Code. 11500914
Brand: Cytiva 80642162
Only null left
Add to basket
Add to basket
Order Info
  • Dimension: 3.54 x 4.13 in. (9 x 10.5cm)
  • Thickness: 0.25 in. (6mm)
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Mini Vertical Gel Electrophoresis Unit
Type Blotting Sponge
Thickness (English) 0.23 in.
Thickness (Metric) 0.6 cm