Learn More
Hirschmann™ Automatic Burettes, Acc. to Dr. Schilling
Product Code. 10350411
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
15 mL
25 mL
50 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
10350411 5 mL Each
10195920 10 mL Each
10569222 15 mL Each
10441331 25 mL Each
10360001 50 mL Each
5 options
Product Code. 10350411
Brand: Hirschmann™ 3600153
Only null left
Add to basket
Add to basket
Excellent for automatically dispensing measured amounts of chemical solutions in the laboratory. Hirschmann Instruments™ Automatic Burettes, Acc. to Dr. Schilling are available in a range of capacities to fit your needs.
  • With Schellbach stripe
  • Blue graduation
  • Fast flow burette
Specifications
Capacity (Metric) 5 mL
Material Glass
Graduations 0.05 mL
Includes LDPE Bottle
Scale Blue
Tolerance ±0.05 mL
Type Schilling Buret
Certifications/Compliance DKD Certificate