Learn More
BRAND™ SILBERBRAND™ AR-GLAS™ Spare Burette
Product Code. 10064310 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
10 mL
15 mL
50 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
10064310 15 mL Each
10238460 10 mL Each
10647671 50 mL Each
3 options
Product Code. 10064310
Brand: BRAND™ 23715
Only null left
Add to basket
Add to basket
Manufactured with Dr. Schilling pattern from AR-GLAS™. For use with Automatic Burettes.
  • AR-GLAS™ construction
  • For use with automatic burette
  • Dr. Schilling pattern
  • Schellbach style back
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 15 mL
Length (Metric) 330 mm
For Use With (Equipment) Automatic burettes
Material Borosilicate Glass
Type Automatic Buret