Learn More
Dräger™ X-act™ 5000 Automatic Tube Pump: Battery Packs
Product Code. p-4910083
Click to view available options
Type:
NiMH Battery Pack
Voltage:
110V
Unit Size:
Each
Product Code. 13547260
Brand: Dräger™ 4523520
Specifications
For Use With (Equipment) X-act 5000 Automatic Tube Pump
Type NiMH Battery Pack
Temperature Range (English) 41°F to 104°F
Temperature Range (Metric) 5° to 40°C
Voltage 110V