Learn More
Dräger™ Short-Term Detector Tubes - Hydrogen Sulfide
Click to view available options
Components Measured:
Hydrogen Sulfide 0.5/a
Hydrogen Sulfide 1/d
Hydrogen Sulfide 100/a
Hydrogen Sulfide 5/b
Measurement Range:
0.5 to 15ppm / 5 to 150ppm
1 to 200ppm
100 to 2000ppm
5 to 60ppm
Unit Size:
Pack of 10
Specifications
Specifications
| Components Measured | Hydrogen Sulfide 0.5/a |
| Humidity Range | 3 to 30mg/L |
| Measurement Range | 0.5 to 15ppm / 5 to 150ppm |
| No. of Strokes | 10 |
| Quantity | 10 |
| Response Time | 6 min. |
| Temperature Range (English) | 32°F to 104°F |
| Temperature Range (Metric) | 0°C to 40°C |